6-Bromo[1,2,4]triazolo[4,3-a]pyridine 97%
Supplier: Ambeed
|
Warning
|
A104857-1G
A104857-25G
A104857-250MG
103796-790EA
16
USD
103796-790
103822-516
77309-376
77623-880
77623-878
6-Bromo[1,2,4]triazolo[4,3-a]pyridine 97%
6-Bromo-[1,2,4]triazolo[4,3-a]pyridine
Precaución: for Research Use Only
Formula:
C₆H₄BrN₃ Melting Pt: 151…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689506 CAS Number: 108281-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...