6-Bromo[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one 95%
Supplier: Ambeed
|
Warning
|
A524308-100MG
A524308-250MG
A524308-5G
A524308-1G
103828-528EA
52
USD
103828-528
76774-664
76774-666
76774-668
77307-722
6-Bromo[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one 95%
6-Bromo[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
Precaución: for Research Use Only
Formula:
C₆H₄BrN₃O Storage Temperature: Ambient |
MDL Number:
MFCD11110678 CAS Number: 425702-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...