6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine 97%

Supplier: Ambeed
Warning

A209273-100G A209273-10G A209273-250MG A209273-1G
103817-928EA 11 USD
103817-928 103848-616 77030-450 77030-452 77030-454 77086-958
6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine 97%
6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₅BrN₄
Storage Temperature: Ambient
MDL Number: MFCD11656619
CAS Number: 947248-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR