6-Benzyl-2,4-dichloro-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A182903-250MG A182903-1G A182903-5G
103815-582EA 28 USD
103815-582 103846-144 76769-612 77301-804 77301-806
6-Benzyl-2,4-dichloro-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine 97%
6-Benzyl-2,4-dichloro-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₄H₁₃Cl₂N₃
Boiling Pt: 423.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08273934
CAS Number: 778574-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR