6-Aminoquinazolin-4(3H)-one 95%

Supplier: Ambeed
Warning

A145794-100MG A145794-250MG A145794-1G
103803-590EA 44 USD
103803-590 103841-670 76767-982 76767-984 77299-474
6-Aminoquinazolin-4(3H)-one 95%
6-Aminoquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₈H₇N₃O
Boiling Pt: 421.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01219542
CAS Number: 17329-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR