6-Aminopyridin-2-ol 98%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxypyridin-2-amine, 6-Aminopyridin-2-ol

A198427-10G A198427-1G A198427-25G
103817-016EA 19 USD
103817-016 103847-642 76647-450 77085-668 77085-670
6-Aminopyridin-2-ol 98%
6-Aminopyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.11 g/mol
Flash Pt: 197 °C
Storage Temperature: Ambient
MDL Number: MFCD00039717
CAS Number: 59315-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR