6-Aminopyridine-2-carbonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 6-Aminopicolinonitrile

A179942-250MG A179942-1G A179942-10G
103807-492EA 12 USD
103807-492 103845-846 77029-390 77083-320 77600-454
6-Aminopyridine-2-carbonitrile 98%
6-Aminopyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.1200 g/mol
Flash Pt: 153.6 °C
Storage Temperature: Ambient
MDL Number: MFCD11044252
CAS Number: 370556-44-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR