6-Aminopicolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Aminopyridine-2-carboxylic acid

A101667-100G A101667-10G A101667-25G A101667-1G A101667-5G
103796-080EA 6 USD
103796-080 103821-776 77107-214 77107-216 77107-218 77282-794 77308-698
6-Aminopicolinic acid 97%
6-Aminopicolinic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
Boiling Pt: 387.8 °C (760 mm Hg)
Melting Pt: 320 °C
Storage Temperature: Ambient
MDL Number: MFCD00233711
CAS Number: 23628-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR