6-Aminoindolin-2-one 97%

Supplier: Ambeed
Warning

A510682-1G A510682-5G A510682-10G A510682-250MG
103828-392EA 93 USD
103828-392 103839-494 76843-480 76843-482 77265-804 77265-806
6-Aminoindolin-2-one 97%
6-Aminoindolin-2-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Boiling Pt: 394.3 °C (760 mm Hg)
Flash Pt: 192.2 °C
Storage Temperature: Ambient
MDL Number: MFCD08669501
CAS Number: 150544-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR