6-Aminoindole 96%

Supplier: Ambeed
Warning

A197028-1G A197028-10G A197028-250MG
103816-888EA 12 USD
103816-888 103847-504 77085-486 77248-752 77248-754
6-Aminoindole 96%
6-Aminoindole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.16
Boiling Pt: 354 °C (760 mm Hg)
Melting Pt: 75…79 °C
Storage Temperature: Freezer
MDL Number: MFCD02093959
CAS Number: 5318-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR