6-Aminohexan-1-ol 98%

Supplier: Ambeed
Warning

A481895-100G A481895-10G A481895-500G A481895-25G A481895-5G
103839-206EA 5 USD
103839-206 76774-368 76774-370 76774-372 76843-148 77307-536
6-Aminohexan-1-ol 98%
6-Aminohexan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
Boiling Pt: 227 °C (760 mm Hg)
Melting Pt: 55…58 °C
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00008241
CAS Number: 4048-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR