(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2-(hydroxymethyl)benzeneboronic acid dehydrate hydrochloride, 6-Aminobenzo[c][1,2]oxaborol-1(3H)-ol hydrochloride
A158832-100MG
A158832-10G
A158832-250MG
A158832-5G
A158832-1G
103805-178EA
149
USD
103805-178
103843-392
77191-004
77191-006
77191-008
77300-372
77319-246
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride 97%
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride
Precaución: Inert atmosphere, 2…8 °C.
Formula:
C₇H₉BClNO₂ MW: 185.42 g/mol Boiling Pt: 484 °C (760 mm Hg) Melting Pt: 300 °C Flash Pt: 246.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04115645 CAS Number: 117098-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...