6-Aminobenzo[b]thiophene 1,1-dioxide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Amino-1,1-dioxobenzo[b]thiophene, 6-Amino-1,1-dioxo-1-benzothiophene
A159861-100MG
A159861-250MG
A159861-5G
A159861-1G
103805-296EA
71
USD
103805-296
103843-522
77235-812
77235-814
77300-424
77319-424
6-Aminobenzo[b]thiophene 1,1-dioxide 97%
6-Aminobenzo[b]thiophene 1,1-dioxide
Precaución: for Research Use Only
Formula:
C₈H₇NO₂S MW: 181.212 g/mol Boiling Pt: 467.4 °C (760 mm Hg) Melting Pt: 150 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00173800 CAS Number: 20503-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...