6-Amino-4-hydroxy-2-mercaptopyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2-thioxo-2,3-dihydropyrimidin-4(1H)-one, 6-Aminothiouracil, 6-Amino-2-thiouracil

A166251-10G A166251-1G A166251-25G A166251-5G A166251-100G
103806-034EA 5 USD
103806-034 103844-294 76872-192 77300-818 77300-820 77320-540 77671-334
6-Amino-4-hydroxy-2-mercaptopyrimidine 98%
6-Amino-4-hydroxy-2-mercaptopyrimidine

Precaución: for Research Use Only

Formula: C₄H₅N₃OS
MW: 143.1670 g/mol
Boiling Pt: 454.7 °C (760 mm Hg)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006077
CAS Number: 1004-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR