6-Amino-4-chloronicotinic acid 97%

Supplier: Ambeed
Warning

A223040-100MG A223040-250MG A223040-50MG A223040-1G A223040-5G
103819-020EA 199 USD
103819-020 103849-898 76814-936 76814-938 76814-940 77303-642 77303-644
6-Amino-4-chloronicotinic acid 97%
6-Amino-4-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
Boiling Pt: 383.1±42.0 °C (760 mm Hg)
Flash Pt: 185.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189221
CAS Number: 1060808-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR