6-Amino-4-((3-chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-7-ethoxyquinoline-3-carbonitrile 95%

Supplier: Ambeed
Warning

A115635-100MG A115635-250MG A115635-1G A115635-5G
103798-598EA 35 USD
103798-598 103824-456 77148-900 77148-902 77297-382 77297-384
6-Amino-4-((3-chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-7-ethoxyquinoline-3-carbonitrile 95%
6-Amino-4-((3-chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-7-ethoxyquinoline-3-carbonitrile

Precaución: for Research Use Only

Formula: C₂₄H₂₀ClN₅O₂
MW: 445.9 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032114
CAS Number: 848139-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR