6-Amino-3-methyluracil 97%

Supplier: Ambeed
Warning

A209391-100MG A209391-10G A209391-250MG A209391-1G A209391-5G A209391-25G
103817-938EA 21 USD
103817-938 103848-624 76975-488 76975-490 76975-492 77086-966 77086-968 77303-096
6-Amino-3-methyluracil 97%
6-Amino-3-methyluracil

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
Melting Pt: 350…352 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02091062
CAS Number: 21236-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR