6-Amino-3,4-dihydroquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A181670-100MG A181670-10G A181670-250MG A181670-1G A181670-5G
103807-636EA 77 USD
103807-636 103846-012 76769-570 76769-572 76769-574 77083-540 77083-542
6-Amino-3,4-dihydroquinolin-2(1H)-one 97%
6-Amino-3,4-dihydroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MW: 162.189 g/mol
Boiling Pt: 423 °C (760 mm Hg)
Flash Pt: 209.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00559318
CAS Number: 22246-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR