6-Amino-3,3-dimethylindolin-2-one 98%

Supplier: Ambeed
Warning

A122684-250MG A122684-1G
103800-256EA 208 USD
103800-256 76766-612 77297-882
6-Amino-3,3-dimethylindolin-2-one 98%
6-Amino-3,3-dimethylindolin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.21 g/mol
Boiling Pt: 366.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11053509
CAS Number: 100510-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR