6-Amino-2H-1,4-benzoxazin-3(4H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-3,4-dihydro-3-oxo-2H-1,4-benzoxazine,6-Amino-3-oxo-3,4-dihydrobenzo[1,4]oxazine,6-Amino-2H-benzo[b][1,4]oxazin-3(4H)-one,6-Amino-4H-benzo[1,4]oxazin-3-one, 6-amino-2,4-dihydro-1,4-benzoxazin-3-one

A259768-100MG A259768-250MG
103812-462EA 25 USD
103812-462 103852-344 77153-416 77153-418
6-Amino-2H-1,4-benzoxazin-3(4H)-one 97%
6-Amino-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O₂
MW: 164.17 g/mol
Melting Pt: 88…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425742
CAS Number: 89976-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR