6-Amino-2-bromo-3-chlorobenzoic acid 95%

Supplier: Ambeed
Warning

A103205-250MG A103205-1G A103205-5G
103796-504EA 87 USD
103796-504 76809-384 77309-086 77548-948
6-Amino-2-bromo-3-chlorobenzoic acid 95%
6-Amino-2-bromo-3-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.477 g/mol
Boiling Pt: 382.4 °C (760 mm Hg)
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD07787451
CAS Number: 65971-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR