6-Amino-2,3-dichlorophenol 97%

Supplier: Ambeed
Warning

A147912-100MG A147912-250MG A147912-1G
103803-848EA 370 USD
103803-848 103841-962 77162-620 77162-622 77299-614
6-Amino-2,3-dichlorophenol 97%
6-Amino-2,3-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20693682
CAS Number: 116278-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR