6-Acetylpicolinonitrile 95%

Supplier: Ambeed
Warning

A200380-100MG A200380-250MG
103817-196EA 39 USD
103817-196 103847-822 77192-258 77192-260
6-Acetylpicolinonitrile 95%
6-Acetylpicolinonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD09263905
CAS Number: 159307-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR