6,8-Dibromo[1,2,4]triazolo[1,5-a]pyrazine 98%

Supplier: Ambeed
Warning

Synonyms: 6,8-Dibromo-[1,2,4]triazolo[1,5-a]pyrazine

A253712-250MG A253712-1G A253712-5G
103812-108EA 28 USD
103812-108 103851-958 77031-610 77304-618 77304-620
6,8-Dibromo[1,2,4]triazolo[1,5-a]pyrazine 98%
6,8-Dibromo[1,2,4]triazolo[1,5-a]pyrazine

Precaución: for Research Use Only

Formula: C₅H₂Br₂N₄
MW: 277.9042 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09701977
CAS Number: 944709-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR