6,7-Dimethoxy-4-hydroxyquinoline 98%

Supplier: Ambeed
Warning

A150721-10G A150721-1G A150721-25G A150721-100G
103804-196EA 6 USD
103804-196 103842-334 76798-642 77299-822 77299-824 77879-047
6,7-Dimethoxy-4-hydroxyquinoline 98%
6,7-Dimethoxy-4-hydroxyquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
Boiling Pt: 355.3 °C (760 mm Hg)
Flash Pt: 168.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07368026
CAS Number: 13425-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR