6,7-Dimethoxy-1-tetralone 97%
Supplier: Ambeed
|
Warning
|
A101690-100MG
A101690-250MG
A101690-1G
A101690-5G
A101690-25G
103796-094EA
28
USD
103796-094
103821-786
76717-566
76717-568
77282-804
77282-806
77878-796
6,7-Dimethoxy-1-tetralone 97%
6,7-Dimethoxy-1-tetralone
Precaución: for Research Use Only
Formula:
C₁₂H₁₄O₃ Boiling Pt: 358.3 °C (760 mm Hg) Melting Pt: 98…100 °C Flash Pt: 161.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134100 CAS Number: 13575-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...