6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A211587-10G
A211587-1G
A211587-25G
A211587-5G
103818-116EA
10
USD
103818-116
103848-810
76800-650
77087-224
77087-226
77087-228
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride 97%
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₄ClNO₂ MW: 227.69 g/mol Melting Pt: 198…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143492 CAS Number: 20232-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...