2,3-Dimethyl-6,7-dimethoxyquinoxaline 98%

Supplier: Ambeed
Warning

Synonyms: 6,7-Dimethoxy-2,3-dimethylquinoxaline

A326517-1G A326517-250MG
103825-438EA 180 USD
103825-438 76839-342 77251-218
2,3-Dimethyl-6,7-dimethoxyquinoxaline 98%
2,3-Dimethyl-6,7-dimethoxyquinoxaline

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MW: 218.252 g/mol
Boiling Pt: 327 °C (760 mm Hg)
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD04038946
CAS Number: 32388-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR