6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline 95%
Supplier: Ambeed
|
Warning
|
A127967-10G
A127967-1G
A127967-25G
A127967-5G
103801-046EA
9
USD
103801-046
76766-986
77313-326
77313-328
77313-330
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline 95%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only
Formula:
C₁₁H₁₅NO₂ Boiling Pt: 314.9 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00777849 CAS Number: 1745-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...