6,7-Dihydropyrazolo[1,5-a]pyridin-4(5H)-one 97%

Supplier: Ambeed
Warning

A116457-10G A116457-250MG A116457-25G A116457-5G A116457-1G
103798-736EA 64 USD
103798-736 103824-602 76983-000 76983-002 76983-004 77297-468 77311-408
6,7-Dihydropyrazolo[1,5-a]pyridin-4(5H)-one 97%
6,7-Dihydropyrazolo[1,5-a]pyridin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 281.8±9.0 °C (760 mm Hg)
Flash Pt: 124.2 °C
Storage Temperature: Ambient
MDL Number: MFCD13176260
CAS Number: 197094-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR