6,7-Dihydroimidazo[1,5-a]pyridin-8(5H)-one 95%
Supplier: Ambeed
|
Warning
|
A198562-100MG
A198562-250MG
A198562-1G
103817-030EA
539
USD
103817-030
103847-658
76825-478
76825-480
77302-590
6,7-Dihydroimidazo[1,5-a]pyridin-8(5H)-one 95%
6,7-Dihydroimidazo[1,5-a]pyridin-8(5H)-one
Precaución: for Research Use Only
Formula:
C₇H₈N₂O MW: 136.15 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11110652 CAS Number: 426219-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...