6,7-Dihydrocyclopenta[b]pyridin-5-one 97%
Supplier: Ambeed
|
Warning
|
A245782-1G
A245782-25G
A245782-5G
A245782-100MG
A245782-250MG
103811-668EA
22
USD
103811-668
103851-498
77090-768
77090-770
77090-772
77165-882
77165-884
6,7-Dihydrocyclopenta[b]pyridin-5-one 97%
6,7-Dihydrocyclopenta[b]pyridin-5-one
Precaución: for Research Use Only
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 247.4 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD07371500 CAS Number: 28566-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...