6,7-Dihydro-5H-pyrido[2,3-b]azepin-8(9H)-one 97%

Supplier: Ambeed
Warning

A119767-100MG A119767-250MG A119767-1G
103799-834EA 307 USD
103799-834 76822-338 76822-340 77311-934
6,7-Dihydro-5H-pyrido[2,3-b]azepin-8(9H)-one 97%
6,7-Dihydro-5H-pyrido[2,3-b]azepin-8(9H)-one

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
Boiling Pt: 314 °C (760 mm Hg)
Melting Pt: 144…145 °C
Flash Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD18253752
CAS Number: 67046-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR