6,7-Difluoro-1H-indazole 97%

Supplier: Ambeed
Warning

A207309-1G A207309-5G A207309-10G A207309-250MG
103817-784EA 36 USD
103817-784 103848-464 77086-736 77086-738 77164-832 77164-834
6,7-Difluoro-1H-indazole 97%
6,7-Difluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂
MW: 154.117 g/mol
Boiling Pt: 281.4±20.0 °C (760 mm Hg)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD18381168
CAS Number: 1260384-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR