2,2'-Methylenebis(6-tert-butyl-4-ethylphenol) 98%

Supplier: Ambeed
Warning

Synonyms: Bis(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methane, 2,2'-Methylenebis(4-ethyl-6-tert-butylphenol), 3,3'-Di-tert-butyl-5,5'-diethyl-2,2'-dihydroxydiphenylmethane, 6,6'-Methylenebis(2-(tert-butyl)-4-ethylphenol)

A108459-100G A108459-25G
103807-996EA 81 USD
103807-996 77245-564 77245-566
2,2'-Methylenebis(6-tert-butyl-4-ethylphenol) 98%
2,2'-Methylenebis(6-tert-butyl-4-ethylphenol)

Precaución: for Research Use Only

Formula: C₂₅H₃₆O₂
MW: 368.56 g/mol
Boiling Pt: 452.1 °C (760 mm Hg)
Melting Pt: 119…122 °C
Flash Pt: 187.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00026296
CAS Number: 88-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR