6,6'-Dimethyl-2,2'-bipyridyl 98%

Supplier: Ambeed
Warning

Synonyms: 6,6'-Dimethyl-2,2'-bipyridine

A307206-10G A307206-250MG A307206-1G A307206-25G A307206-5G A307206-100G
103814-494EA 8 USD
103814-494 103835-670 76772-740 76772-742 77305-950 77305-952 77305-954 77601-384
6,6'-Dimethyl-2,2'-bipyridyl 98%
6,6'-Dimethyl-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MW: 184.2371 g/mol
Boiling Pt: 278.6 °C (760 mm Hg)
Melting Pt: 93…95 °C
Flash Pt: 104.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00059776
CAS Number: 4411-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR