6,10-Dioxaspiro[4.5]decane-7,9-dione 97%

Supplier: Ambeed
Warning

A250671-100MG A250671-250MG A250671-5G A250671-1G
103811-940EA 80 USD
103811-940 103851-778 77031-512 77031-514 77091-136 77304-536
6,10-Dioxaspiro[4.5]decane-7,9-dione 97%
6,10-Dioxaspiro[4.5]decane-7,9-dione

Precaución: for Research Use Only

Formula: C₈H₁₀O₄
MW: 170.163 g/mol
Boiling Pt: 444.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01617888
CAS Number: 58093-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR