6-(Trifluoromethyl)pyrimidin-4-amine 98%

Supplier: Ambeed
Warning

A163770-250MG A163770-1G A163770-100MG
103805-764EA 31 USD
103805-764 103844-026 77138-252 77300-676 77685-338
6-(Trifluoromethyl)pyrimidin-4-amine 98%
6-(Trifluoromethyl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C5H4F3N3
Storage Temperature: Refrigerator
MDL Number: MFCD08436657
CAS Number: 672-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR