3-Amino-6-(trifluoromethyl)pyridazine 95%

Supplier: Ambeed
Warning

Synonyms: 6-(Trifluoromethyl)pyridazin-3-amine

A180201-250MG A180201-1G
103807-522EA 142 USD
103807-522 103845-876 76985-734 77301-666
3-Amino-6-(trifluoromethyl)pyridazine 95%
3-Amino-6-(trifluoromethyl)pyridazine

Precaución: for Research Use Only

Formula: C₅H₄F₃N₃
MW: 163.101 g/mol
Boiling Pt: 297.7±40.0 °C (760 mm Hg)
Flash Pt: 133.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189559
CAS Number: 935777-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR