6-(Trifluoromethyl)nicotinic acid 98%

Supplier: Ambeed
Warning

A110849-100G A110849-10G A110849-25G
103797-740EA 5 USD
103797-740 103823-548 77136-020 77136-022 77136-024
6-(Trifluoromethyl)nicotinic acid 98%
6-(Trifluoromethyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₂
Boiling Pt: 259.3 °C (760 mm Hg)
Melting Pt: 193…197 °C
Flash Pt: 110.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00792430
CAS Number: 231291-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR