6-(Trifluoromethyl)indole 98%

Supplier: Ambeed
Warning

A481059-1G A481059-5G A481059-10G A481059-250MG
103828-124EA 29 USD
103828-124 103839-198 76843-142 76843-144 77279-478 77279-480
6-(Trifluoromethyl)indole 98%
6-(Trifluoromethyl)indole

Precaución: for Research Use Only

Formula: C₉H₆F₃N
MW: 185.15 g/mol
Boiling Pt: 256.7 °C (760 mm Hg)
Melting Pt: 101 °C
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00272316
CAS Number: 13544-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR