6-(Trifluoromethyl)-1H-indazole 97%

Supplier: Ambeed
Warning

A150803-100MG A150803-250MG A150803-1G A150803-5G
103804-202EA 46 USD
103804-202 103842-340 77010-740 77010-742 77317-870 77317-872
6-(Trifluoromethyl)-1H-indazole 97%
6-(Trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
Boiling Pt: 321.5±42.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08690097
CAS Number: 954239-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR