6-Hydroxymethylpyridine-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Hydroxymethyl)picolinonitrile

A399368-100MG A399368-250MG A399368-1G A399368-5G
103827-030EA 110 USD
103827-030 103838-068 76773-702 76773-704 76841-506 76841-508
6-Hydroxymethylpyridine-2-carbonitrile 97%
6-Hydroxymethylpyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.135 g/mol
Boiling Pt: 310.5 °C (760 mm Hg)
Flash Pt: 141.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07367926
CAS Number: 50501-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR