6-(Hydroxymethyl)picolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Hydroxymethyl)pyridine-2-carboxylic acid

A124882-250MG A124882-1G A124882-5G
103800-590EA 57 USD
103800-590 76766-842 77312-732 77312-734
6-(Hydroxymethyl)picolinic acid 97%
6-(Hydroxymethyl)picolinic acid

Precaución: for Research Use Only

Formula: C₁H₁NO₃
MW: 153.1354 g/mol
Boiling Pt: 381.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09032021
CAS Number: 1197-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR