2-(Fluoromethyl)-6-formylpyridine 95%

Supplier: Ambeed
Danger

Synonyms: 6-(Fluoromethyl)picolinaldehyde

A131550-250MG A131550-1G
103801-600EA 162 USD
103801-600 76811-206 77314-098
2-(Fluoromethyl)-6-formylpyridine 95%
2-(Fluoromethyl)-6-formylpyridine

Precaución: for Research Use Only

Formula: C₇H₆FNO
MW: 139.1300 g/mol
Boiling Pt: 214.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11865234
CAS Number: 208111-28-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR