6-(Ethoxycarbonyl)pyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Ethoxycarbonyl)picolinic acid

A134777-10G A134777-250MG A134777-1G A134777-5G
103802-048EA 16 USD
103802-048 76798-000 76798-002 77314-818 77314-820
6-(Ethoxycarbonyl)pyridine-2-carboxylic acid 97%
6-(Ethoxycarbonyl)pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.172 g/mol
Boiling Pt: 396.2 °C (760 mm Hg)
Melting Pt: 116…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00806154
CAS Number: 21855-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR