6-(Difluoromethoxy)pyridin-3-amine 97%

Supplier: Ambeed
Warning

A328890-100MG A328890-250MG
103825-502EA 120 USD
103825-502 103836-456 76989-238 77612-892
6-(Difluoromethoxy)pyridin-3-amine 97%
6-(Difluoromethoxy)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆F₂N₂O
MW: 160.121 g/mol
Boiling Pt: 249.8±35.0 °C at N/A mmHg
Flash Pt: 104.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11847318
CAS Number: 317810-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR