6-(Difluoromethoxy)nicotinic acid ≥95%

Supplier: Ambeed
Warning

A143932-100MG A143932-250MG
103803-350EA 475 USD
103803-350 77889-956 77889-957
6-(Difluoromethoxy)nicotinic acid ≥95%
6-(Difluoromethoxy)nicotinic acid

Precaución: Sealed in dry, at room temperature.

Formula: C₇H₅F₂NO₃
Boiling Pt: 276.7±35.0 °C (760 mm Hg)
Flash Pt: 121.2 °C
Storage Temperature: Ambient
MDL Number: MFCD18258473
CAS Number: 1211535-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR