6-(Difluoromethoxy)nicotinaldehyde 98%

Supplier: Ambeed
Warning

A245323-100MG A245323-250MG A245323-5G A245323-1G
103811-636EA 131 USD
103811-636 76987-758 76987-760 76987-762 77304-356
6-(Difluoromethoxy)nicotinaldehyde 98%
6-(Difluoromethoxy)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.117 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18261899
CAS Number: 1211577-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR