6'-(Diethylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one 98%

Supplier: Ambeed
Warning

Synonyms: 7-Anilino-3-diethylamino-6-methyl fluoran

A213133-25G A213133-100G A213133-500G
103818-262EA 9 USD
103818-262 103848-964 77087-444 77261-116 77261-118
6'-(Diethylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one 98%
6'-(Diethylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

Precaución: for Research Use Only

Formula: C₃₁H₂₈N₂O₃
Boiling Pt: 673.9±55.0 °C (760 mm Hg)
Melting Pt: 195…197 °C
Flash Pt: 361.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00603523
CAS Number: 29512-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR